Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST2575

Lead paint film for portable x-ray fluorescence analyzers

NIST® SRM® 2575, nominal 0.3 mg/cm2 (color code:  green)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 1 +blank

manufacturer/tradename

NIST®

technique(s)

XRF: suitable

application(s)

coatings

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

Each SRM 2575 unit contains a white polyester sheet about 7.6 cm in width, 10.2 cm in length, and 0.2 mm thick. This sheet is coated with a uniform layer of green-colored paint, measuring approximately 0.04 mm in thickness.



SRM 2575_cert

SRM 2575_SDS

Application

Leadpaint film for building surfaces reference material (SRM) is intended to validate the results obtained from portable, hand-held, X-ray fluorescence analyzers while conducting lead testing in paint coatings on exterior and interior building surfaces.

Features and Benefits

  • Available with a certificate documenting NIST-certified values of total lead areic mass in cured paint and the values are based on measurements by isotope dilution inductively coupled plasma mass spectrometry(ID-ICP-MS).
  • Certified values are metrological traceability is to the SI units for mass and length (expressed as milligrams percentimeter-squared).
  • The NIST certificate is provided with expiration certificate, storage, handling,use, and maintenance instructions.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Lead (Pb)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service