Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST610

Trace elements in glass

NIST® SRM® 610

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 4 wafers

manufacturer/tradename

NIST®

application(s)

glass & ceramic

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 610 contains four wafers of an oval to circular cross-section and a nominal diameter ranging from 12 mm (about 0.47 in) to 14 mm (about 0.55 in) with 3 mm thickness, sliced from a hand-pulled rod.

SRM 610_cert

SRM 610_SDS

Application

The (SRM) is used to develop chemical analysis methods for trace elements in glass.

Features and Benefits

  • Available with a certificate documenting NIST-certified mass fraction values of constituents in Trace elements in glass.
  • The reference and information values of elements and isotoperatio are presented in mass fractions.
  • The NIST certificate is provided with expiration certificate, storage, handling, use,and maintenance instructions.

Other Notes

Certified for the analytes listed below.
Antimony (Sb), Arsenic (As), Barium (Ba), Cadmium (Cd), Chromium (Cr), Iron (Fe), Lead (Pb), Manganese (Mn), Nickel (Ni), Rubidium (Rb), Selenium (Se), Silver (Ag), Strontium (Sr), Thorium (Th), Uranium (U)

Reference/Informational value is provided for the following analytes.
Copper (Cu), Thallium (Tl), Strontium (87Sr/86Sr)
Boron (B), Cobalt (Co), Gold (Au), Lithium (Li), Potassium (K), Titanium (Ti), Zinc (Zn), Uranium (235U/UTotal)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service