MilliporeSigma
F1149
Carestream® X-Omat LS film
Synonym(s):
autoradiography film, xray film
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.22
UNSPSC Code:
41105341
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
size
8 in. (19.5 cm) × 10 in. (25.5 cm)
General description
Kodak Life Science Imaging film is equivalent to XRP. Coated with emulsion on both sides of a blue-tinted base, produces higher quality images than XAR film (improved definition with less background fog), however, the exposure time is approximately doubled. Sheets of film are interleaved with sheets of paper.
Legal Information
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Still not finding the right product?
Explore all of our products under Carestream® X-Omat LS film
Choose from one of the most recent versions:
Certificates of Analysis (COA)