Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST1577C

Bovine liver

NIST® SRM® 1577c

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

solid

packaging

pkg of 20 g

manufacturer/tradename

NIST®

technique(s)

mass spectrometry (MS): suitable

format

matrix material

storage temp.

10-30°C

General description

This bovine liver standard reference material (SRM) consists of liver tissues from healthy steers. The tissue is collected and prepared under strict protocols designed to preserve the original composition and minimize contamination. A unit of NIST1577C consists of one bottle containing 20 g of freeze-dried liver powder.

NIST1577C_Cert

NIST1577C_SDS

Application

The bovine liver standard reference material (SRM) is intended for use in evaluating the accuracy of analytical methods for 25-hydroxyvitamin D3 in animal tissues and other biological materials.

Other Notes

  • Details on expiry, storage, usage, and sample preparation and analysis are available in the NIST certificate.
  • Information on disposal and transportation is documented in SDS.
Certified for the analytes listed below. See certificate for values and more details
Nitrogen, Phosphorous
Trace Elements: Antimony (SB), Arsenic (As), Cadmium (Cd), Calcium (Ca), Cesium (Cs), Chlorine (Cl2), Chromium (Cr), Cobalt (Co), Copper (Cu), Gold (Au), Hydrogen (H), Iron (Fe), Lead (Pb), Lithium (Li), Magnesium (Mg), Manganese (Mn), Mercury (Hg), Molybdenum (Mo), Nickel (Ni), Potassium (K), Rubidium (Rb), Selenium (Se), Silicium (Si), Sodium (Na), Strontium (Sr), Sulfur (S), Vanadium (V), Zinc (Zn)

Matrix Group: Meat

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michał Majewski et al.
Pharmacological reports : PR, 71(5), 862-869 (2019-08-14)
Experimental studies have emphasized that cardiovascular alterations can be improved by the long-term use of resveratrol (trans-3,5,4'-trihydroxystilbene; RSV) as well as dietary copper (Cu) intake. Male Wistar rats were supplemented for 8 weeks with Cu (6.5 mg/kg diet) as either nanoparticles
Validation of a microwave-assisted digestion procedure of pate samples using diluted HNO3 for Fe and Zn determination by FS FAAS
Silveria N. P. S, et al.
Analytical Methods : Advancing Methods and Applications, 5, 6411-6411 (2013)
Jianzong Zhou et al.
Journal of agricultural and food chemistry, 70(15), 4765-4773 (2022-04-07)
Quantification of trace and minor nutrient elements is crucial for maintaining human health. A reliable laser ablation inductively coupled plasma mass spectrometry (LA-ICP-MS) method for food materials was developed by combining fine food powder (dm < 3 μm) pellets as
Michał Majewski et al.
Toxicology and applied pharmacology, 390, 114877-114877 (2020-01-10)
Copper (Cu) deficiency plays an important role in the development of cardiovascular disorders. Resveratrol (RSV) possesses pleiotropic cardiovascular benefits; however, the mechanism(s) by which RSV exerts protective effects are not completely understood. Male Wistar rats at 6 weeks of age were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service