Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P4347

Sigma-Aldrich

Propylene Glycol

meets USP testing specifications

Synonym(s):

1,2-Propanediol, 1,2-PDO, Methylethyl glycol, Propane-1,2-diol, Propylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1340498
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
application(s):
general analytical
pharmaceutical (small molecule)
sample preparation
bp:
187 °C (lit.)
vapor pressure:
0.08 mmHg ( 20 °C)

Agency

USP/NF
meets USP testing specifications

Quality Level

vapor density

2.62 (vs air)

vapor pressure

0.08 mmHg ( 20 °C)

autoignition temp.

779 °F

expl. lim.

12.5 %

refractive index

n20/D 1.432 (lit.)

bp

187 °C (lit.)

mp

−60 °C (lit.)

density

1.036 g/mL at 25 °C (lit.)

application(s)

general analytical
pharmaceutical (small molecule)
sample preparation

SMILES string

CC(O)CO

InChI

1S/C3H8O2/c1-3(5)2-4/h3-5H,2H2,1H3

InChI key

DNIAPMSPPWPWGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propylene glycol, a low-molecular-weight aliphatic alcohol, holds significance as a key solvent in biological and pharmaceutical formulation research. Its outstanding miscibility with water and diverse organic compounds, coupled with its low freezing point, low vapor pressure, and non-toxic nature, positions it as a preferred choice for a variety of research applications, particularly in sensitive biological assays and pharmaceutical preparations.

Going beyond its solvent capabilities, propylene glycol excels as an excipient in pharmaceutical formulations, enhancing the solubility and stability of therapeutic molecules and pharmaceutical additives. This attribute proves indispensable in excipient and drug delivery research, contributing to the development of more efficacious and stable medications. In biochemical and physiological research, propylene glycol functions as a potent cryoprotectant, safeguarding cells and tissues from freezing-induced damage. Its application extends to investigations of drug absorption, metabolism, and distribution, providing researchers with a crucial tool to decipher the intricate journey of drugs within the body.

The versatility of propylene glycol extends to molecular biology, particularly in Polymerase Chain Reaction (PCR), where it safeguards DNA and RNA strands, ensuring reliable and reproducible results. Studies demonstrate its exceptional ability to stabilize DNA for extended periods, making it an essential component for long-term sample preservation in genetic and genomic analyses. In the pharmaceutical industry, propylene glycol finds extensive use as a solvent and extraction medium, playing a pivotal role in drug development research, facilitating efficient extraction and purification of bioactive compounds.

Application

Propylene glycol has been used:
  • for removing excess Oil Red O stain in zebrafish larvae
  • in the preparation of alginate hydrogels
  • for the dissolution of androstenone
  • for the calibration of cone beam scanner in computed tomography

Features and Benefits

  • Versatile and adaptable for a wide variety of laboratory and research applications
  • Meets USP testing specifications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

comparable product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-Chia Tseng et al.
Disease models & mechanisms, 11(9) (2018-08-24)
Niemann-Pick disease type C1 (NPC1) is a rare autosomal recessive lysosomal storage disease primarily caused by mutations in NPC1 NPC1 is characterized by abnormal accumulation of unesterified cholesterol and glycolipids in late endosomes and lysosomes. Common signs include neonatal jaundice
Li-Hao Huang et al.
Cell metabolism, 29(2), 475-487 (2018-11-13)
Lipoproteins trapped in arteries drive atherosclerosis. Extravascular low-density lipoprotein undergoes receptor uptake, whereas high-density lipoprotein (HDL) interacts with cells to acquire cholesterol and then recirculates to plasma. We developed photoactivatable apoA-I to understand how HDL passage through tissue is regulated.
Shawn C Chafe et al.
Cancer immunology research, 7(7), 1064-1078 (2019-05-16)
Treatment strategies involving immune-checkpoint blockade (ICB) have significantly improved survival for a subset of patients across a broad spectrum of advanced solid cancers. Despite this, considerable room for improving response rates remains. The tumor microenvironment (TME) is a hurdle to
Burak Altun et al.
Ultrasonics, 110, 106274-106274 (2020-11-02)
Tissue-mimicking materials (TMMs) play a key role in the quality assurance of ultrasound diagnostic equipment and should have acoustic properties similar to human tissues. We propose a method to quantify the acoustic properties of TMM samples through the use of
R S Rajasenan et al.
Toxicology and applied pharmacology, 135(1), 89-99 (1995-11-01)
Engrafting components of human immune systems in severe combined immunodeficient (SCID) mice has been utilized to investigate the pathogenesis of several human autoimmune diseases and may provide a model for studying idiosyncratic drug toxicity. The purpose of this investigation was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service