Skip to Content
MilliporeSigma

356350

Glycerol

greener alternative

Synonym(s):

Glycerin, 1,2,3-Propanetriol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
NACRES:
NA.25
UNSPSC Code:
50161901
Beilstein/REAXYS Number:
635685
MDL number:
Assay:
≥99% (GC)
Technique(s):
electrophoresis: suitable
Bp:
182 °C/20 mmHg (lit.)
Vapor pressure:
<1 mmHg ( 20 °C)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

3.1 (vs air)

Quality Segment

vapor pressure

<1 mmHg ( 20 °C)

description

Merck USA index - 14, 4484

assay

≥99% (GC)

form

liquid

autoignition temp.

698 °F

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated

greener alternative product characteristics

Safer Solvents and Auxiliaries
Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

electrophoresis: suitable

color

clear colorless

refractive index

n20/D 1.474 (lit.)

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

density

1.25 g/mL (lit.)

greener alternative category

shipped in

ambient

storage temp.

10-30°C

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

General description

Obtained from oils and fats as a biproduct in the manufacturing of soaps and fatty acids.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of the 12 Principles of Green Chemistry. Glycerol, with its high boiling point, low flammability, and polarity, is a standout choice. As a non-toxic and biodegradable compound, it aligns perfectly with the principles of ′Safer Solvents and Auxiliaries′ and ′Design for Degradation′. Click here for more information.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


Still not finding the right product?

Explore all of our products under Glycerol


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

390.2 °F - Pensky-Martens closed cup

flash_point_c

199 °C - Pensky-Martens closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library