Skip to Content
MilliporeSigma
All Photos(7)

Documents

791784

Sigma-Aldrich

Tetrabutylammonium hydrogensulfate

anhydrous, free-flowing, Redi-Dri, 97%

Synonym(s):

Tetrabutylammonium bisulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(HSO4)
CAS Number:
Molecular Weight:
339.53
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

product line

Redi-Dri

Assay

97%

form

crystals

quality

free-flowing

mp

169-171 °C (lit.)

SMILES string

OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.H2O4S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

SHFJWMWCIHQNCP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetrabutylammonium bisulfate(TBAHS), a quaternary ammonium salt, is a phase-transfer reagent. It severs as cocatalyst, additive, acid, also as ion-paring agent.

Application

Tetrabutylammonium hydrogensulfate is used as a cocatalyst in the synthesis of propionic acid and the ketones by hydrocarboxylation of ethylene.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

348.1 °F - closed cup

Flash Point(C)

175.6 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haji Bahadar et al.
Environmental toxicology and pharmacology, 39(3), 1161-1169 (2015-05-04)
Benzene (C6H6) is an organic compound used in petrochemicals and numerous other industries. It is abundantly released to our environment as a chemical pollutant causing widespread human exposure. This study mainly focused on benzene induced toxicity on rat pancreatic islets
K M Frankowski et al.
Journal of dairy science, 97(9), 5356-5370 (2014-07-16)
Many food companies are trying to limit the amount of sodium in their products. Permeate, the liquid remaining after whey or milk is ultrafiltered, has been suggested as a salt substitute. The objective of this study was to determine the
Hiroyuki Nakao et al.
Biochimica et biophysica acta, 1848(1 Pt A), 145-150 (2014-10-15)
KcsA is a pH-dependent potassium channel that is activated at acidic pH. The channel undergoes global conformational changes upon activation. We hypothesized that the open-close conformational changes of the transmem brane region could promote the flip-flop of phosphoiipids. Based on
Muhammet Uyanik et al.
Science (New York, N.Y.), 345(6194), 291-294 (2014-07-19)
The diverse biological activities of tocopherols and their analogs have inspired considerable interest in the development of routes for their efficient asymmetric synthesis. Here, we report that chiral ammonium hypoiodite salts catalyze highly chemo- and enantioselective oxidative cyclization of γ-(2-hydroxyphenyl)ketones
Jia Bao et al.
Environmental science and pollution research international, 21(12), 7650-7655 (2014-03-14)
Severe perfluoroalkyl acid (PFAA) contamination resulting from the fast-growing semiconductor, electrochemical, and optoelectronic industries has been determined in the river water in the vicinity of the Taipei area, Taiwan, during recent years. However, little is known about body burdens of

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service