Skip to Content
MilliporeSigma
Tetrabutylammonium hydrogensulfate anhydrous, free-flowing, Redi-Dri™, 97%

Tetrabutylammonium hydrogensulfate

Tetrabutylammonium hydrogensulfate anhydrous, free-flowing, Redi-Dri™, 97%
Synonym(s):
Tetrabutylammonium bisulfate
Linear Formula:
(CH3CH2CH2CH2)4N(HSO4)
CAS Number:
Molecular Weight:
339.53
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2