MilliporeSigma
  • pH-dependent promotion of phospholipid flip-flop by the KcsA potassium channel.
pH-dependent promotion of phospholipid flip-flop by the KcsA potassium channel.
Biochimica et biophysica acta (2014-10-15)
Hiroyuki Nakao, Keisuke Ikeda, Masayuki Iwamoto, Hirofumi Shimizu, Shigetoshi Oiki, Yasushi Ishihama, Minoru Nakano
ABSTRACT
KcsA is a pH-dependent potassium channel that is activated at acidic pH. The channel undergoes global conformational changes upon activation. We hypothesized that the open-close conformational changes of the transmem brane region could promote the flip-flop of phosphoiipids. Based on this hypothesis, we measured the flip-flop ofNBD-labeled phospholipids in KcsA-incorporated proteoliposomes. Both flip and flop rates of ~NBD-PC were significantly enhanced in the presence of KcsA and were several times higher at pH 4.0 than at pH 7.4, suggesting that KcsA promotes the phospholipid flip in a conformation-dependent manner. Phospholipids were nonselectively flipped with respect to the glycerophospholipid structure. In the active state of KcsA channel,tetrabutylammonium locks the channel in the open conformation at acidic pH; however, it did not alter the fliprate of CGNBD-PC. Thus, the open-close transition of the transmembrane region did not affect the flip-flop of phospholipids. In addition, the KcsA mutant that lacked anN-terminal amphipathic helix (MO-helix) was found to show reduced ability to fl ip C6NBD-phospholipids at acidic pH. The closed conformation is stabilized in the absence of MO-heli x, and thus the attenuated flip could be explained by the reduced prevalence of the open conformation.These results suggest that the open conformation of KcsA can disturb the bilayer integrity and facilitate the flip-flop of phospholipids.
Sigma-Aldrich
Tetrabutylammonium cyanide, technical, ≥80%
Sigma-Aldrich
2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, ≥99.0% (TLC)
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic solution, 1.0 M in H2O
Sigma-Aldrich
2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, ≥95.5% (GC), ≥98% (TLC)
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, 97%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 53.5-56.5% in H2O
Sigma-Aldrich
Tetrabutylammonium cyanide, 95%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
Supelco
Tetrabutylammonium hydroxide solution, ~40% in water, suitable for ion chromatography
Sigma-Aldrich
Tetrabutylammonium perchlorate, ≥95.0% (T)
Supelco
Tetrabutylammonium perchlorate, for electrochemical analysis, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, anhydrous, free-flowing, Redi-Dri, 97%
Supelco
Tetrabutylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Tetrabutylammonium bisulfate solution, suitable for ion pair chromatography, LiChropur, concentrate, ampule
Supelco
Tetrabutylammonium iodide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium bromide solution, 50 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium azide
Supelco
Tetrabutylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium iodide, reagent grade, 98%
Sigma-Aldrich
Tetrabutylammonium chloride, ≥97.0% (NT)
Sigma-Aldrich
4-Chloro-7-nitrobenzofurazan, BioReagent, suitable for fluorescence, ≥97.0% (HPLC)
Sigma-Aldrich
Tetrabutylammonium iodide, ≥99.0% (AT)
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Supelco
Tetrabutylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium nitrate, 97%
Sigma-Aldrich
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic, puriss., 99% (T)