Skip to Content
MilliporeSigma

1.18312

Tetrabutylammonium hydrogen sulfate

for ion pair chromatography LiChropur™

Synonym(s):

Tetra-n-butylammonium hydrogen sulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H37NO4S
CAS Number:
Molecular Weight:
339.53
UNSPSC Code:
12352107
EC Index Number:
251-068-5
NACRES:
NB.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Tetra-n-butylammonium hydrogen sulfate, for ion pair chromatography LiChropur™

Quality Segment

assay

≥99% (acidimetric)

technique(s)

HPLC: suitable, LC/MS: suitable, ion chromatography: suitable, ion pair chromatography: suitable

loss

≤2 loss on drying, 120°C, vacuum

transmittance

200 nm, ≥70%, 220 nm, ≥90%, 250 nm, ≥98%, (0.005 M; 1 cm in H2O)

pH

≤2 (20 °C, 100 g/L in H2O)

mp

168-174 °C

bulk density

650 kg/m3

suitability

passes test for identity (IR)

storage temp.

2-30°C

SMILES string

[S](=O)(=O)([O-])[O-].[N+](CCCC)(CCCC)(CCCC)CCCC.[H+]

InChI

1S/C16H36N.H2O4S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

SHFJWMWCIHQNCP-UHFFFAOYSA-M

General description

Tetra-n-butylammoniumhydrogen sulfate is commonly used as an ion-pair reagent and ion interactionreagent (IIR) in chromatographic analysis.

Application

Tetra-n-butylammonium hydrogen sulfate can be used as an:
  • Ion interaction reagent (IIR) in an RP-HPLC method for the determination of acetic acid (AA) and trifluoroacetic acid (TFA)in synthetic peptides.
  • Ion pair reagent in the IPE (Ion Pair Extraction) method.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1

WGK

WGK 2

Flash Point (°F)

348.1 °F - closed cup

Flash Point (°C)

175.6 °C - closed cup

Hazard Codes

Storage Class

8A - Combustible corrosive hazardous materials



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library