Skip to Content
MilliporeSigma

Skip To

795488

Potassium phosphate monobasic

anhydrous, free-flowing, Redi-Dri™, ACS reagent, ≥99%

Synonym(s):

Potassium dihydrogen phosphate, prim.-Potassium phosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
KH2PO4
CAS Number:
Molecular Weight:
136.09
NACRES:
NA.21
PubChem Substance ID:
UNSPSC Code:
12352302
EC Number:
231-913-4
MDL number:
Assay:
≥99%
Grade:
ACS reagent, anhydrous
Form:
powder or crystals
Solubility:
water: soluble (208 g/l at 20 °C (68 °F))
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

ACS reagent, anhydrous

product line

Redi-Dri™

assay

≥99%

form

powder or crystals

quality

free-flowing

impurities

≤0.01% Insoluble matter

loss

≤0.2% loss on drying, 105°C

pH

4.1-4.5 (25 °C, 5%)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

mp

252.6 °C (lit.)

solubility

water: soluble (208 g/l at 20 °C (68 °F))

density

2.338 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%, sulfate (SO42-): ≤0.003%

cation traces

Fe: ≤0.002%, Na: ≤0.005%, heavy metals: ≤0.001% (by ICP)

SMILES string

[K+].OP(O)([O-])=O

InChI

1S/K.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

GNSKLFRGEWLPPA-UHFFFAOYSA-M

General description

Potassium phosphate monobasic (monopotassium orthophosphate) is a water soluble inorganic salt. Its standard enthalpy of formation has been reported to be −376.1 kcalmol-1. It acts a starting material in the synthesis of potassium metaphosphate.

Redi-Dri™ technology eliminates clumping and the need for complicated procedures to keep your salts free-flowing. Keeping your salts dry helps improve your results by giving accurate measurements and easier-to-handle salts and buffers. Rather than drying your salts in an oven or desiccator or breaking them apart manually, Redi-Dri™ ensures that your salts will remain easy-to-use for months without worrying about anti-caking agents. Even after repeated openings you can be assured that your results are always accurate. Redi-Dri™ saves you time, energy and money by keeping your salts usable for longer!

Application

Potassium phosphate monobasic has been used in the preparation of phosphate buffer and phosphate buffered saline (PBS). It has also been used in the preparation of PEG1000 (polyethylene glycol)/potassium phosphate aqueous two-phase systems (ATPS).

Features and Benefits

  • Increased efficiency – Eliminate time and effort of loosening hardened powders
  • Safety – Avoid risks of using tools of force to break up clumps
  • Economy – Faster preparation and solvation increase productivity and reduce costs
  • Assured quality – Excellent, expert-tested quality with no anti-caking agents
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RDD020
description

anhydrous, free-flowing, Redi-Dri™, ACS reagent, ≥99%

description

anhydrous, free-flowing, Redi-Dri™, ReagentPlus®, ≥99.0% (titration)

form

powder or crystals

form

crystalline

solubility

water: soluble (208 g/l at 20 °C (68 °F))

solubility

water: 100 mg/mL, clear, colorless

assay

≥99%

assay

≥99.0% (titration)

Quality Level

200

Quality Level

100

grade

anhydrous, ACS reagent

grade

anhydrous

anion traces

sulfate (SO42-): ≤0.003%, chloride (Cl-): ≤0.001%

anion traces

-


Still not finding the right product?

Explore all of our products under Potassium phosphate monobasic


Storage Class

11 - Combustible Solids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–5 of 5 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How long will material remain free-flowing after it is opened?

    1 answer
    1. While the exact amount of time your material will remain free-flowing can vary, this largely depends on the ambient storage conditions and how many times it is opened.

      Helpful?

  4. How does Redi-Dri™ help keep salts and buffers free-flowing?

    1 answer
    1. Redi-Dri™ products are specially handled and the bottle is specially designed to ensure that the product will remain dry without adding impurities or additives to the product. Ensuring that you receive the highest quality material that is easy to handle.

      Helpful?

  5. Are there any special storage conditions that must be maintained?

    1 answer
    1. No, Redi-Dri™ products can be stored under the same conditions as the equivalent non-Redi-Dri™ products.

      Helpful?

Reviews

No rating value

Active Filters