Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

72548

Millipore

Nitrate Broth

NutriSelect® Plus, suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

sterility

non-sterile

Quality Level

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

composition

meat extract, 3 g/L
peptone, 5 g/L
potassium nitrate, 1 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | metabolite detection: suitable
microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

application(s)

agriculture
environmental
food and beverages
water monitoring

microbiology

suitability

anaerobic bacteria
nonselective for Bacillus spp.
nonselective for Enterobacter spp.
nonselective for Enterococcus spp.
nonselective for Escherichia coli
nonselective for Klebsiella spp.
nonselective for Neisseria spp.
nonselective for Pseudomonas spp.
nonselective for Salmonella spp.
nonselective for Serratia spp.
nonselective for Shigella spp.
nonselective for Staphylococcus spp.
nonselective for coliforms

General description

Nitrate broth is used for the nitrate test. It can be used for bacteria like Enterobacteriaceae, Bacilli and Salmonella.

Application

for detecting nitrate-reducing and indole-producig microorganisms

Preparation Note

Dissolve 9 g in 1 litre distilled water. Sterilize by autoclaving at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circle

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aybike Kamiloğlu et al.
Brazilian journal of microbiology : [publication of the Brazilian Society for Microbiology], 51(3), 1279-1287 (2020-04-28)
Five Lactobacillus strains isolated from sucuk (Turkish dry-fermented sausage) were studied for their genetic and technological properties. For genotypic identification, strains 16S rRNA gene sequences were used. To determine the antimicrobial activity of strains, seven foodborne pathogens were tested. Strains

Articles

An article concerning the detection, identification, differentiation, and cultivation of Pseudomonas species.

An article concerning the detection, identification, differentiation, and cultivation of Pseudomonas species.

An article concerning the detection, identification, differentiation, and cultivation of Pseudomonas species.

An article concerning the detection, identification, differentiation, and cultivation of Pseudomonas species.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service