Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M1428

Millipore

MIO Medium

NutriSelect® Plus, suitable for microbiology

Synonym(s):

Motility Indole Ornithine Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

sterility

non-sterile

Quality Level

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | growth & motility: suitable
microbe id | specific enzyme detection: suitable
microbiological culture: suitable

final pH

6.5±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages
veterinary

microbiology

suitability

Escherichia coli
nonselective and differential for Enterobacter spp.
nonselective and differential for Klebsiella spp.
nonselective and differential for Proteus spp.
nonselective and differential for enterobacteriaceae

Application

Used for the identification of enterobacteriaceae on the basis of motility, indole production and ornithine decarboxylase activity.

Components

Ingredients (g/L)
Casein enzymatic hydrolysate, 10.00
Peptic digest of animal tissue, 10.00
Yeast extract, 3.00
L-Ornithine hydrochloride, 5.00
Dextrose, 1.00
Bromocresol purple, 0.02
Agar, 2.00

Preparation Note

Suspend 31 g of MIO Medium in 1000 ml of distilled water. Boil to dissolve the medium completely. Dispense 5 ml into each test tube. Sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes. Cool the tubes in an upright position.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service