Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51138

Millipore

Nitrate Reagent Disks Kit

suitable for microbiology, to detect an microorganism’s ability to reduce nitrate

Synonym(s):

Nitrate reduction test

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

disc size

6 mm

Quality Level

sterility

sterile

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 50 pieces

technique(s)

microbe id | metabolite detection: suitable

application(s)

agriculture
clinical testing
environmental
food and beverages
pharmaceutical

microbiology

suitability

Enterobacter spp.
Neisseria spp.
anaerobic bacteria
bacteria

General description

Nitrate Reagent Disks Kit is used to detect an organism’s ability to reduce nitrate. The test involves the detection of the enzyme nitrate reductase, which causes the reduction of nitrate in the presence of a suitable electron donor to nitrite, which can be tested by an appropriate colorimetric reagent. Almost all Enterobacteriaceae reduce nitrate. The nitrate in the disc is broken down to nitrite when nitrate reductase positive culture is grown in presence of these discs. The reduction of nitrate (NO3) to nitrite (NO2) and subsequently to nitrogen gas (N2) usually takes place under anaerobic conditions, in which an organism derives its oxygen from nitrate. Most facultative anaerobes can reduce nitrate in the absence of oxygen.

Application

Nitrate Reagent Disks Kit is used for rapid detection and confirmation of an organism’s ability to reduce nitrate. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Components

Sterile filter paper discs (50pcs, diameter 6mm) impregnated with Nitrate reagent (Cat. No. 08086).
Rehydrating fluids (Cat. No. 43505, 5ml)

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service