Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75315

Millipore

OF Test Nutrient Agar

suitable for microbiology, NutriSelect® Basic

Synonym(s):

Oxidative Fermentative Agar acc. to Hugh a. Leifson

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.74

sterility

non-sterile

Quality Level

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 2.5 g/L
bromothymol blue, 0.08 g/L
casein peptone (pancreatic), 2 g/L
dipotassium hydrogen phosphate, 0.3 g/L
sodium chloride, 5 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbe id | utilization test: suitable
microbiological culture: suitable

final pH

7.1±0.2 (25 °C)

antibiotic activity spectrum

Gram-negative bacteria

application(s)

agriculture
clinical testing
environmental
food and beverages

microbiology

suitability

Alcaligenes spp.
Chromobacterium spp.
Moraxella spp.
Pasteurella spp.
Acinetobacter spp.
Aeromonas spp.
flavobacteria
nonselective and differential for Bordetella spp.
nonselective and differential for Campylobacter spp.
nonselective and differential for Enterococcus spp.
nonselective and differential for Escherichia coli
nonselective and differential for Pseudomonas spp.
nonselective and differential for Salmonella spp.
nonselective and differential for Shigella spp.
nonselective and differential for Vibrio spp.
nonselective and differential for Yersinia spp.
nonselective and differential for bacteria (General Media)
nonselective and differential for coliforms
nonselective and differential for enterobacteriaceae

Application

Medium for demonstrating oxidative and fermentative catabolism of carbohydrate acc. to Hugh and Leifson (1953). In particular for the differentiation and classification of gram-negative intestinal bacteria.

Preparation Note

Dissolve 9.88 g in 1 litre distilled water. Sterilize by autoclaving at 121°C for 15 minutes. Let cool to approx. 50°C and add 100 ml of sterile filtered 10% D(+)-glucose (Cat. No. 49140), lactose (Cat. No. 61340), sucrose (Cat. No. 84100) or other carbohydrate.

Other Notes

Studies on coagulase-positive staphylococci from bovine milk

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The taxonomic significance of fermentative versus oxidative metabolism of carbohydrates by various gram negative bacteria.
R HUGH et al.
Journal of bacteriology, 66(1), 24-26 (1953-07-01)
J A Erasmus
The Onderstepoort journal of veterinary research, 52(1), 25-29 (1985-03-01)
When conventional techniques were applied to 84 isolates of Staphylococcus aureus from milk samples, it was found that they were all catalase and phosphatase positive and oxidase negative. They all fermented glucose within 24 h and mannitol within 24-48 h

Articles

Vibrios are motile, curved or comma-shaped bacilli and have a single polar flagella with sheet proteins. They are often found in open water, freshwater and saltwater.

Specific agars and broths support detection and cultivation of Campylobacter, requiring complex media with essential supplements.

Specific agars and broths support detection and cultivation of Campylobacter, requiring complex media with essential supplements.

Specific agars and broths support detection and cultivation of Campylobacter, requiring complex media with essential supplements.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service