Skip to Content
MilliporeSigma
All Photos(1)

Documents

53477

Millipore

CFC Selective Supplement

ready-to-use, for Pseudomonas spp., suitable for microbiology

Synonym(s):

Pseudomonas Selective Supplement, Cetrimide Fucidin Cephaloridine Selective Supplement

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41171614
NACRES:
NA.85

Agency

according to ISO 13720:2010

Quality Level

sterility

sterile

form

powder

shelf life

limited shelf life, expiry date on the label

feature

ready-to-use

packaging

pkg of 5 ampules

application(s)

clinical testing
environmental
food and beverages
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

Pseudomonas spp.

General description

The CFC Selective Supplement is a microbiological medium supplement designed for the selective isolation of Pseudomonas species. The supplement facilitates the growth of desired Pseudomonas species while inhibiting others. Each vial of supplement is sufficient for preparing 500 mL of medium.

Application

CFC Selective Supplement is used for the selective isolation of Pseudomonas species in a variety of testing environments, including clinical, environmental, food and beverage, and pharmaceutical testing.

Components

per vial sufficient for 500 mL medium
Cetrimide 5mg, Fucidin 5mg, Cephaloridine 25mg

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Gastrointestinal tract

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service