Skip to Content
MilliporeSigma
All Photos(1)

Documents

P2102

Millipore

Pseudomonas agar base

suitable for microbiology, NutriSelect® Plus

Synonym(s):

CFC Pseudomonas agar, base, CN-agar, Cetrimide nalidixic acid agar, base, PP Pseudomonas agar, base

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

Agency

according to ISO 11059:2009
according to ISO 13720:2010
according to ISO 16266:2006

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.1±0.2 (25 °C)

application(s)

agriculture
bioburden testing
cleaning products
cosmetics
environmental
food and beverages
personal care
water monitoring

microbiology

suitability

selective for Pseudomonas spp.

Application

Pseudomonas agar base may be used for the selective detection of Pseudomonas aeruginosa pathogens in mineral water sample that are mainly responsible for food and waterborne infections.

Components

Ingredients (g/L)
Peptic digest of animal tissue, 16.00
Casein enzymic hyrolysate, 10.00
Potassium sulphate, 10.00
Magnesium chloride • 6H2O, 1.40
Agar, 11.00

Preparation Note

Suspend 24.2 g of Pseudomonas Agar Base in 500 ml of distilled water containing 5 ml of glycerol Cat. No. 49767). Heat to boiling to dissolve the medium completely. Sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes. Cool to 50°C and aseptically add the sterile rehydrated contents of either Cetrinix Supplement (C8721) or C.F.C. Supplement (Cat. No. 53477). Mix well and pour into sterile petri plates.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Modified Pseudomonas agar: new differential medium for the detection/enumeration of Pseudomonas aeruginosa in mineral water.
Ramalho R, et al.
Journal of Microbiological Methods, 49(1), 69-74 (2002)
Han-Gyu Park et al.
Scientific reports, 7(1), 2168-2168 (2017-05-21)
Climate change causes permafrost thawing, and we are confronted with the unpredictable risk of newly discovered permafrost microbes that have disease-causing capabilities. Here, we first characterized the detailed chemical structure of the lipid A moiety from a Pseudomonas species that

Articles

An article concerning the detection, identification, differentiation, and cultivation of Pseudomonas species.

An article concerning the detection, identification, differentiation, and cultivation of Pseudomonas species.

An article concerning the detection, identification, differentiation, and cultivation of Pseudomonas species.

An article concerning the detection, identification, differentiation, and cultivation of Pseudomonas species.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service