Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.05284

Millipore

Cetrimide agar

Pseudomonas selective agar base for microbiology (According harm. EP/USP/JP)

Synonym(s):

Pseudomonas selective agar, CETRIMIDE agar, Pseudomonas selective agar (base)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

EP
JP
USP

Quality Level

sterility

non-sterile

form

granular

manufacturer/tradename

GranuCult® prime

technique(s)

microbiological culture: suitable

pH

7.0-7.4 (37 °C, 45 g/L in H2O, after autoclaving)

solubility

45 g/L

bulk density

640 kg/m3

application(s)

microbiology
pharmaceutical

storage temp.

15-25°C

suitability

Pseudomonas spp.

General description

Cetrimide agar is recommended for the selective isolation and identification of Pseudomonas aeruginosa. The use of cetrimide (N-Cetyl-N,N,N-trimethyl ammonium bromide) largely inhibits the growth of the accompanying microbial flora minimizes interference with the growth of P. aeruginosa. This medium does not inhibit the pigment production of Pseudomonas aeruginosa.

Application

Cetrimide agar (or Pseudomonas selective agar) is used for the isolation and differentiation of Pseudomonas aeruginosa from pharmaceutical products and various other materials.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs. The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service