Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MHS32

Sigma-Aldrich

Mayer′s Hematoxylin Solution

Synonym(s):

(6aS,11bR)-7,11b-Dihydroindeno[2,1-c]chromene-3,4,6a,9,10(6H)-pentol, Delafield′s hematoxylin, Progressive stains

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116124
PubChem Substance ID:
NACRES:
NA.47

form

solution

Quality Level

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

1 g/L

technique(s)

microbe id | staining: suitable

pH

2.4 (25 °C)

application(s)

hematology
histology

storage temp.

room temp

SMILES string

Oc1cc2C[C@@]3(O)COc4c(O)c(O)ccc4[C@H]3c2cc1O

InChI

1S/C16H14O6/c17-10-2-1-8-13-9-4-12(19)11(18)3-7(9)5-16(13,21)6-22-15(8)14(10)20/h1-4,13,17-21H,5-6H2/t13-,16+/m0/s1

InChI key

WZUVPPKBWHMQCE-XJKSGUPXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Used as a counterstain for procedures such as immunohistochemistry or laser microdissection.

Application

General purpose nuclear stain, progressive type. Used with hematoxylin and eosin staining.

Other Notes

1 g/L certified hematoxylin

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ludivine C Litzler et al.
Nature communications, 10(1), 22-22 (2019-01-04)
Mechanisms regulating B cell development, activation, education in the germinal center (GC) and differentiation, underpin the humoral immune response. Protein arginine methyltransferase 5 (Prmt5), which catalyzes most symmetric dimethyl arginine protein modifications, is overexpressed in B cell lymphomas but its
J Verney et al.
The journal of nutrition, health & aging, 21(10), 1133-1141 (2017-12-01)
Effect of 3 different dairy protein sources on the recovery of muscle function after limb immobilization in old rats. Longitudinal animal study. Institut National de la Recherche Agronomique (INRA). The study took part in a laboratory setting. Old rats were
Maud Creze et al.
Muscle & nerve, 56(4), 787-796 (2016-12-23)
The ultrastructure of a nerve has implications for surgical nerve repair. The aim of our study was to characterize the fascicular versus fibrillar anatomy and the autonomic versus somatic nature of the fetal sciatic nerve (SN). Immunohistochemistry for vesicular acetylcholine
Abdelbasset Hamrouni et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 25(10), 3104-3114 (2019-02-28)
Mycosis fungoides is one of the most common types of extranodal T-cell lymphomas, considered to be caused by malignant transformation of the mature T cells residing in the skin. However, some clinical observations such as the multifocal distribution of mycosis
Sebastian Dütting et al.
Nature communications, 8, 15838-15838 (2017-06-24)
Blood platelets are produced by large bone marrow (BM) precursor cells, megakaryocytes (MKs), which extend cytoplasmic protrusions (proplatelets) into BM sinusoids. The molecular cues that control MK polarization towards sinusoids and limit transendothelial crossing to proplatelets remain unknown. Here, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service