Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.52098

Sigma-Aldrich

Fmoc-Glu(EDANS)-OH

≥95.0% (HPLC), for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-Glu(EDANS)-OH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H31N3O8S
CAS Number:
Molecular Weight:
617.67
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

Product Name

Fmoc-Glu(EDANS)-OH, Novabiochem®

Quality Level

product line

Novabiochem®

Assay

≥95% (TLC)
≥95.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

15-25°C

SMILES string

[S](=O)(=O)(O)c1c2c(c(ccc2)NCCNC(=O)CC[C@H](NC(=O)OCC3c4c(cccc4)c5c3cccc5)C(=O)O)ccc1

InChI

1S/C32H31N3O8S/c36-30(34-18-17-33-27-13-5-12-25-24(27)11-6-14-29(25)44(40,41)42)16-15-28(31(37)38)35-32(39)43-19-26-22-9-3-1-7-20(22)21-8-2-4-10-23(21)26/h1-14,26,28,33H,15-19H2,(H,34,36)(H,35,39)(H,37,38)(H,40,41,42)/t28-/m0/s1

InChI key

HPYLYODKLGEPPX-NDEPHWFRSA-N

General description

Fluorescence-labeled amino acid for preparing fluorescence-quenched peptide substrates by Fmoc SPPS [1]. Most frequently used in conjunction with Dabcyl quenching group.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Chromogenic Labeling of Synthetic Peptides

Literature references

[1] J. W. Drijfhout, et al. in ′Peptides, Chemistry, Structure & Biology, Proc. 14th American Peptide Symposium′, P. T. P. Kaumaya & R. S. Hodges (Eds), Kingswinford, Mayflower Scientific Ltd., 1996, pp. 129.

Linkage

Replaces: 04-12-1238

Analysis Note

Colour (visual): slight grey to grey
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.0 % (a/a)
Purity (TLC(CMA1)): ≥ 95 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Chromogenic and fluorogenic derivatives are invaluable tools for biochemistry, having numerous applications in enzymology, protein chemistry, immunology and histochemistry.

Chromogenic and fluorogenic derivatives are invaluable tools for biochemistry, having numerous applications in enzymology, protein chemistry, immunology and histochemistry.

Chromogenic and fluorogenic derivatives are invaluable tools for biochemistry, having numerous applications in enzymology, protein chemistry, immunology and histochemistry.

Chromogenic and fluorogenic derivatives are invaluable tools for biochemistry, having numerous applications in enzymology, protein chemistry, immunology and histochemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service