Skip to Content
MilliporeSigma

8.55054

EDANS NovaTag®_resin

Novabiochem®

Synonym(s):

EDANS-MPB-AM resin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.22
UNSPSC Code:
12352116
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

NovaSyn® TG, Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

15-25°C

General description

The EDANS/dabcyl fluorophore-quencher pair is one of the most commonly-used for FRET peptides, owing to excellent spectral overlap between the emission spectrum of EDANS and absorbance spectrum of dabcyl [1]. The introduction of the EDANS moiety is traditionally achieved either by coupling of a peptide fragment to EDANS in solution [1, 2] or through the use of a pre-formed derivative, such as Fmoc-Glu(EDANS)-OH [3]. Novabiochem′s EDANS resin enables for the first time the direct synthesis of C-terminally EDANS-labeled peptides by solid phase synthesis [4]. Loading of the first residue requires acylation of the resin-bound secondary amine and is, therefore, best carried out using HATU activation. Chain extension and cleavage can then be effected using standard methods. EDANS NovaTag resin has been recently employed to prepare fluorescently labeled aminoalkane diphenyl phosphonate affinity probes for chymotrypsin- and elastase-like serine proteases [5].

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins Literature references

[1] G. T. Wang, et al. (1990) Tetrahedron Lett., 31, 6493.
[2] C. Garcia-Echeverria & D. H. Rich (1992) Febs Lett., 297, 100.
[3] J. W. Drijfhout, et al. in ′Peptides, Chemistry, Structure& Biology, Proc. 14th American Peptide Symposium′, P. T. P. Kaumaya &R. S. Hodges (Eds), Kingswinford, Mayflower Scientific Ltd., 1996, pp. 129.
[4] J. Beythien & P. White (2005) Tetrahedron Lett., 46,101.
[5] B. F. Gilmore, et al. (2009) Bioconjugate Chem., 20, 2098.

Analysis Note

Color (visual): white to yellow to brown
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.30 - 0.60 mmol/g
Swelling Volume (in DMF): lot specific result
The polymer matrix is copoly styrene - 1 & DVB 100 - 200 mesh

Other Notes

Replaces: 04-12-3904

Legal Information

NOVASYN is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVATAG is a registered trademark of Merck KGaA, Darmstadt, Germany
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library