Skip to Content
MilliporeSigma

8.51022

Dabcyl-OSu

Novabiochem®

Synonym(s):

Dabcyl-OSu, N-(4-[4Æ-(Dimethylamino)phenylazo]benzoyloxy)succinimide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H18N4O4
CAS Number:
Molecular Weight:
366.37
UNSPSC Code:
12352108
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥98.0% (HPLC)

form

powder

reaction suitability

reagent type: fluorescent-labelling reagent

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

15-25°C

SMILES string

N3(C(=O)CCC3=O)OC(=O)c1ccc(cc1)N=Nc2ccc(cc2)N(C)C

InChI

1S/C19H18N4O4/c1-22(2)16-9-7-15(8-10-16)21-20-14-5-3-13(4-6-14)19(26)27-23-17(24)11-12-18(23)25/h3-10H,11-12H2,1-2H3

InChI key

IBOVDNBDQHYNJI-UHFFFAOYSA-N

General description

Pre-activated derivative for the introduction of the Dabcyl group during SPPS. The Dabcyl group quenches the fluorescence of EDANS, Mca, TET, JOE, FAM fluorophores, making it an extremely useful tool for the synthesis of fluorescence-quenched peptide substrates.

Associated Protocols and Technical Articles
Chromogenic Labeling of Synthetic Peptides

Analysis Note

Color (visual): red
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(011A)): ≥ 98 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 01-63-0105

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library