Skip to Content
MilliporeSigma

8.51037

Fmoc-NH-(PEG)-COOH (9 atoms)

≥99.0% (HPLC), for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-NH-(PEG)-COOH (9 atoms), Fmoc-8-amino-3,6-dioxaoctanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H23NO6
CAS Number:
Molecular Weight:
385.41
UNSPSC Code:
12352106
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fmoc-NH-(PEG)-COOH (9 atoms), Novabiochem®

Quality Segment

product line

Novabiochem®

assay

≥98% (TLC), ≥99.0% (HPLC)

form

powder

reaction suitability

reaction type: Pegylations

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

N(CCOCCOCC(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C21H23NO6/c23-20(24)14-27-12-11-26-10-9-22-21(25)28-13-19-17-7-3-1-5-15(17)16-6-2-4-8-18(16)19/h1-8,19H,9-14H2,(H,22,25)(H,23,24)

InChI key

XQPYRJIMPDBGRW-UHFFFAOYSA-N

General description

9-Atom long aminoacid PEG spacer. Can be introduced using standard Fmoc SPPS protocols

Application

Fmoc-NH-(PEG)-COOH (9 atoms) Novabiochem®. CAS 166108-71-0, molar mass 385.42 g/mol.

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Purity (TLC(011A)): ≥ 98 %
Assay (HPLC, area%): ≥ 99.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 01-63-0203

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library