Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.52099

Sigma-Aldrich

Fmoc-Lys(Dnp)-OH

Novabiochem®

Synonym(s):

Fmoc-Lys(Dnp)-OH, N-α-Fmoc-N-ε-2,4-dinitrophenyl-L-lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H26N4O8
CAS Number:
Molecular Weight:
534.52
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥97.0% (HPLC)
≥98% (TLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

15-25°C

SMILES string

[N+](=O)([O-])c1c(ccc(c1)[N+](=O)[O-])NCCCC[C@H](NC(=O)OCC2c3c(cccc3)c4c2cccc4)C(=O)O

InChI

1S/C27H26N4O8/c32-26(33)24(11-5-6-14-28-23-13-12-17(30(35)36)15-25(23)31(37)38)29-27(34)39-16-22-20-9-3-1-7-18(20)19-8-2-4-10-21(19)22/h1-4,7-10,12-13,15,22,24,28H,5-6,11,14,16H2,(H,29,34)(H,32,33)/t24-/m0/s1

InChI key

OENCMORJQAUAAJ-DEOSSOPVSA-N

General description

A modified lysine derivative for the preparation of chromogenically-labeled peptides by Fmoc SPPS.The Dnp group is the preferred quencher for use in conjunction with the Mca fluorophore, making it an extremely useful tool for the synthesis of fluorescence-quenched peptide substrates.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Chromogenic Labeling of Synthetic Peptides

Linkage

Replaces: 04-12-1239

Analysis Note

Color (visual): slight yellow to yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in DMF): -13.5 - -10.5 °
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Assay (HPLC, area%): ≥ 97.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service