Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.51036

Sigma-Aldrich

Fmoc-NH-(PEG)₄-COOH (19 atoms)

Novabiochem®

Synonym(s):

Fmoc-NH-(PEG)₄-COOH (19 atoms), Fmoc-18-amino-4,7,10,13,16-pentaoxaoctadecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H37NO9
CAS Number:
Molecular Weight:
531.59
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥97% (TLC)
≥97.0% (HPLC)

form

oil

reaction suitability

reaction type: Pegylations

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

N(CCOCCOCCOCCOCCOCCC(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C28H37NO9/c30-27(31)9-11-33-13-15-35-17-19-37-20-18-36-16-14-34-12-10-29-28(32)38-21-26-24-7-3-1-5-22(24)23-6-2-4-8-25(23)26/h1-8,26H,9-21H2,(H,29,32)(H,30,31)

InChI key

TWQTXZPTZPOEEB-UHFFFAOYSA-N

General description

19-Atom long aminoacid PEG spacer. Can be introduced using standard Fmoc SPPS protocols

Linkage

Replaces: 01-63-0200

Analysis Note

Colour (visual): pale yellow to beige
Appearance of substance (visual): oil
Identity (LC-MS): 530.2 - 532.2
Purity (TLC(011A)): ≥ 97 %
Assay (HPLC, area%): ≥ 97.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service