Skip to Content
MilliporeSigma

8.51034

Fmoc-NH-(PEG)2-COOH (13 atoms)

Novabiochem®

Synonym(s):

Fmoc-NH-(PEG)₂-COOH (13 atoms), Fmoc-12-amino-4,7,10-trioxadodecanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H29NO7
CAS Number:
Molecular Weight:
443.49
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥98% (TLC), ≥99.0% (HPLC)

form

solid

reaction suitability

reaction type: Pegylations

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

N(CCOCCOCCOCCC(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C24H29NO7/c26-23(27)9-11-29-13-15-31-16-14-30-12-10-25-24(28)32-17-22-20-7-3-1-5-18(20)19-6-2-4-8-21(19)22/h1-8,22H,9-17H2,(H,25,28)(H,26,27)

InChI key

CHIDDYZONKDHLG-UHFFFAOYSA-N

General description

13-Atom long aminoacid PEG spacer. Can be introduced using standard Fmoc SPPS protocols

Application

Fmoc-NH-(PEG)₂-COOH (13 atoms) Novabiochem®. CAS 867062-95-1, molar mass 443.5 g/mol.

Analysis Note

Color (visual): white to slight yellow to light beige
Appearance (visual): powder or oil
Identity (LC-MS): 442.5 - 444.5
Purity (TLC(011A)): ≥ 98 %
Assay (HPLC, area%): ≥ 99.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.
The product may be a white powder or a transparent glass-like solid/oil

Other Notes

Replaces: 01-63-0198

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library