Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

86407

Sigma-Aldrich

TentaGel S RAM

extent of labeling: ~0.24 mmol/g loading

Synonym(s):

N-Fmoc-4′-[poly(oxyethylene)carbamoylmethoxy]-2,4-dimethoxybenzhydrylamine polymer-bound, Poly(oxyethylene)-RAM polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

product line

TentaGel

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

~0.24 mmol/g loading

particle size

~90 μm

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

General description

TentaGel S RAM is a Fmoc protected resin and modified with a Rink amide linker, suitable for solid-phase peptide synthesis.

Application

TentaGel S RAM is used in the solid-phase synthesis of aryl ethers by Mitsunobu reaction. Additionally, it can be used in Suzuki-coupling reaction of organoboronic acids with iodo- and bromo-substituted benzoic acid.

Legal Information

TentaGel is a trademark of Rapp Polymere GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid phase synthesis of aryl ethers via the Mitsunobu reaction
Tetrahedron Letters, 36, 3789-3792 (1995)
Rapid microwave-assisted Suzuki coupling on solid-phase
Tetrahedron Letters, 37, 8219-8222 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service