Skip to Content
MilliporeSigma

8.51038

Fmoc-NH-(PEG)5-COOH (22 atoms)

≥95% (TLC), for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-NH-(PEG)5-COOH (22 atoms), Fmoc-21-amino-4,7,10,13,16,19-hexaoxaheneicosanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C30H41NO10
CAS Number:
Molecular Weight:
575.65
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fmoc-NH-(PEG)5-COOH (22 atoms), Novabiochem®

Quality Segment

product line

Novabiochem®

assay

≥93.0% (HPLC), ≥95% (TLC)

form

oil

reaction suitability

reaction type: Pegylations

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

N(CCOCCOCCOCCOCCOCCOCCC(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C30H41NO10/c32-29(33)9-11-35-13-15-37-17-19-39-21-22-40-20-18-38-16-14-36-12-10-31-30(34)41-23-28-26-7-3-1-5-24(26)25-6-2-4-8-27(25)28/h1-8,28H,9-23H2,(H,31,34)(H,32,33)

InChI key

HEGZERUHBVYZPH-UHFFFAOYSA-N

General description

22-Atom long aminoacid PEG spacer. Can be introduced using standard Fmoc SPPS protocols

Application

Fmoc-NH-(PEG)5-COOH (22 atoms) Novabiochem®. CAS 882847-34-9, molar mass 575.61 g/mol.

Analysis Note

Color (visual): colourless to slight yellow to beige
Appearance of substance (visual): oil
Identity (IR): passes test
Purity (TLC(011A)): ≥ 95 %
Assay (HPLC, area%): ≥ 93.0 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 01-63-0204

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library