Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.41811

Sigma-Aldrich

Copper(I) cyanide

for synthesis

Synonym(s):

Copper(I) cyanide, Cupricin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CCuN
CAS Number:
Molecular Weight:
89.56
MDL number:
UNSPSC Code:
12352302
EC Index Number:
208-883-6
NACRES:
NA.22
grade:
for synthesis

grade

for synthesis

Quality Level

mp

473 °C

density

2.92 g/cm3

bulk density

750 kg/m3

storage temp.

2-30°C

SMILES string

[Cu+].N#[C-]

InChI

1S/CN.Cu/c1-2;/q-1;+1

InChI key

DOBRDRYODQBAMW-UHFFFAOYSA-N

Application

  • Catalytic reduction of nitroaromatic compounds: Copper nanoparticles produced via pulsed laser ablation of Copper(I) cyanide in different solvents show significant catalytic activity for the reduction of toxic nitroaromatic compounds, highlighting its utility in environmental remediation (Begildayeva et al., 2021).
  • Synthesis and spectroscopy of boranes: Research details the facile synthesis of B-Cyanodicarba-closo-dodecaboranes using Copper(I) cyanide, exploring its implications for spectroscopic features and potential applications in boron chemistry (Wingen and Scholz, 2016).

Analysis Note

Assay of Cu: ≥ 70.5 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service