Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

216305

Sigma-Aldrich

Copper(I) cyanide

99%

Synonym(s):

Cuprous cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuCN
CAS Number:
Molecular Weight:
89.56
Beilstein:
3587244
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

Assay

99%

form

powder

mp

474 °C (lit.)

density

2.92 g/mL at 25 °C (lit.)

SMILES string

[Cu]C#N

InChI

1S/CN.Cu/c1-2;

InChI key

DULSAGLWMRMKCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Applied in a widely useful, thiosulfate-assisted, synthesis of a variety of diamine-CuCN complexes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Liver,spleen,Bone marrow

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Virender K Sharma et al.
Environmental science & technology, 39(10), 3849-3854 (2005-06-15)
Copper(Il) cyanide (Cu(CN)4(3-)) in the gold mine industry presentsthe biggest concern in cyanide management because it is much more stable than free cyanide. Cu(CN)4(3-) is highlytoxic to aquatic life; therefore, environmentally friendly techniques are required for the removal of Cu(CN)4(3-)
Fred B. Stocker et al.
Inorganic chemistry, 35(11), 3145-3153 (1996-05-22)
The syntheses and crystal structures of the first cyanide, sulfur mixed ligand copper(I) complexes are reported. The first complex of the family was discovered when (CuCN)(3)(C(6)H(12)N(4))(2) (1) (C(6)H(12)N(4) = hexamethylenetetramine) was treated with aqueous thiourea. The sulfur ligands include thiourea
Douglas B Grotjahn et al.
Journal of the American Chemical Society, 124(20), 5895-5901 (2002-05-16)
Copper(I) cyanide is an important reagent in organic, organometallic, and supramolecular chemistry because of both the copper center and the versatile cyanide ligand. Solid-phase CuCN and many of its derivatives show oligomeric or polymeric structures, a trait shared by other
Fred B. Stocker et al.
Inorganic chemistry, 38(5), 984-991 (2001-10-24)
A new synthetic procedure developed recently in our laboratories has made possible the synthesis of variety of new complexes of CuCN with diamines. Synthesis was effected by adding the ligand to a solution of CuCN in aqueous sodium thiosulfate. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service