Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

184535

Sigma-Aldrich

Silver cyanide

99%

Synonym(s):

Argentous cyanide, Silver monocyanide 2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgCN
CAS Number:
Molecular Weight:
133.89
Beilstein:
4360924
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

powder

SMILES string

[Ag]C#N

InChI

1S/CN.Ag/c1-2;

InChI key

MFIHOCAEOJNSOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in preparation of Tl[Ag(CN)2], which exhibits previously unknown Ag(CN)2 luminescence.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad A. Omary et al.
Inorganic chemistry, 37(6), 1380-1386 (2001-10-24)
The structure of thallium dicyanoargentate(I) has been determined crystallographically. The crystal structure shows an Ag-Ag distance of 3.11 Å. This is the shortest Ag-Ag distance reported for any silver dicyanide salt whose crystal structure has been determined. Raman spectra of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service