Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.41022

Sigma-Aldrich

n-Dodecyltrimethylammonium bromide

for synthesis

Synonym(s):

n-Dodecyltrimethylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H34BrN
CAS Number:
Molecular Weight:
308.34
MDL number:
UNSPSC Code:
12352107
EC Index Number:
214-290-3
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<1 hPa ( 20 °C)

Quality Level

form

powder

potency

200-1000 mg/kg LD50, oral (Rat)

mp

217 °C

solubility

>954 g/L

density

1.17 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Br-].[N+](CCCCCCCCCCCC)(C)(C)C

InChI

1S/C15H34N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16(2,3)4;/h5-15H2,1-4H3;1H/q+1;/p-1

InChI key

XJWSAJYUBXQQDR-UHFFFAOYSA-M

Application

  • Surfactant Applications in Oil Recovery: Research demonstrates the utility of n-Dodecyltrimethylammonium bromide in enhancing oil recovery under high-salinity, high-temperature conditions. The surfactant showed remarkable long-term thermal stability, relevant for industrial applications in oil fields (Hou et al., 2024).
  • Dye Adsorption Studies: The interaction of n-Dodecyltrimethylammonium bromide with alginate hydrogel beads was studied for the removal of Malachite Green dye. This research highlighted the impact of surfactant hydrophobicity on adsorption efficiency, presenting a potential for wastewater treatment technologies (Malik et al., 2024).
  • Role in Precipitation of Reactive Dyes: A study explored the effect of alkyl chain length in surfactants like n-Dodecyltrimethylammonium bromide on the precipitation behavior of Reactive Brilliant Blue KN-R, suggesting potential applications in textile manufacturing processes (Liu et al., 2024).
  • Thermoresponsive Self-assembly in Nanoscale Systems: n-Dodecyltrimethylammonium bromide was utilized to study the generation of thermoresponsive self-assemblies in Tetronics®, providing insights into the temperature-dependent behaviors of surfactant mixtures at the nanoscale (Patel et al., 2024).
  • Electrostatic Adsorption in Surfactant Mixtures: This research described the electrostatically driven adsorption behaviors observed in mixed surfactant systems including n-Dodecyltrimethylammonium bromide, pertinent to applications in coatings and formulations requiring surface activity modifications (Vilangottunjalil et al., 2024).

Analysis Note

Assay (argentometric): ≥ 97.0 %
Identity (IR): passes test

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service