Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

1.04727

Supelco

Isooctane

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

2,2,4-Trimethylpentane, 2,2,4-Trimethylpentane, Isobutyltrimethylmethane, iso-Octane, Isooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2C(CH3)3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696876
MDL number:
UNSPSC Code:
12191502
EC Index Number:
208-759-1
NACRES:
NB.21
grade:
ACS reagent
for analytical purposes
Assay:
≥99.5% (GC)
bp:
98-99 °C (lit.)
vapor pressure:
41 mmHg ( 21 °C)
Pricing and availability is not currently available.

grade

ACS reagent
for analytical purposes

Quality Level

Agency

reag. Ph. Eur.

vapor density

3.9 (vs air)

vapor pressure

41 mmHg ( 21 °C)

product line

EMSURE®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

745 °F

potency

>2500 mg/kg LD50, oral (Rat)

expl. lim.

6 %

impurities

≤0.0003 meq/g Acidity
≤0.005% Sulfur compounds (as S)
≤0.01% Water

evapn. residue

≤0.001%

color

APHA: ≤10

transmittance

250-420 nm, ≥98%

refractive index

n20/D 1.391 (lit.)

pH

7

bp

98-99 °C (lit.)

mp

−107 °C (lit.)

transition temp

flash point -12 °C

density

0.692 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%

storage temp.

2-30°C

SMILES string

CC(C)CC(C)(C)C

InChI

1S/C8H18/c1-7(2)6-8(3,4)5/h7H,6H2,1-5H3

InChI key

NHTMVDHEPJAVLT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Behavior of acetyl modified amino acids in reverse micelles: a non-invasive and physiochemical approach.: This study utilizes isooctane in reverse micelles to explore the behavior of acetyl modified amino acids. The findings emphasize isooctane′s role in facilitating non-invasive and physiochemical analyses, highlighting its importance in analytical chemistry and biochemical studies (Mehta et al., 2007).

  • Nonionic surfactants: a key to enhance the enzyme activity at cationic reverse micellar interface.: The research investigates the use of isooctane in forming cationic reverse micelles with nonionic surfactants to enhance enzyme activity. Isooctane′s effectiveness in this context underscores its value in biocatalysis and enzymatic studies (Shome et al., 2007).

  • Lecithin organogels used as bioactive compounds carriers. A microdomain properties investigation.: This study explores the application of isooctane in lecithin organogels for carrying bioactive compounds. Isooctane′s role in stabilizing the microdomains within the organogels is crucial for their effectiveness in drug delivery systems (Avramiotis et al., 2007).

  • Switching electrical conductivity in an AOT-isooctane-water microemulsion through photodimerization of solubilized N-methyl-2-quinolone.: This research demonstrates the use of isooctane in AOT-water microemulsions to switch electrical conductivity via photodimerization. The study highlights isooctane′s utility in developing responsive materials for advanced chemical and physical applications (Bufe and Wolff, 2006).

  • Photochromism of crown ethers with incorporated azobenzene moiety.: Isooctane is utilized in studying the photochromism of crown ethers with azobenzene moieties. The findings underscore isooctane′s role in facilitating the investigation of photoresponsive materials, relevant to materials science and photochemistry (Janus and Sworakowski, 2005).

Linkage

Discover EMSURE - to be more than sure!

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service