Skip to Content
MilliporeSigma

Skip To

1.01226

Ammonium monovanadate

GR for analysis Reag. Ph Eur

Synonym(s):

Ammonium monovanadate, Ammonium metavanadate, Ammonium vanadate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
H4NO3V
CAS Number:
Molecular Weight:
116.98
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-261-3
NACRES:
NB.21
Assay:
≥99%
Form:
solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

reag. Ph. Eur.

Quality Segment

assay

≥99%

form

solid

potency

169 mg/kg LD50, oral (Rat), >2500 mg/kg LD50, skin (Rat)

pH

7 (20 °C, 5.1 g/L in H2O)

mp

200 °C (decomposition)

solubility

5.1 g/L

density

2.3 g/cm3 at 20 °C

bulk density

1000 kg/m3

storage temp.

2-30°C

SMILES string

[V](=O)(=O)O.N

InChI

1S/H3N.H2O.2O.V/h1H3;1H2;;;/q;;;;+1/p-1

InChI key

YBVKNHXQSRDWAA-UHFFFAOYSA-M

Application


  • Femtosecond laser self-assembly for silver vanadium oxide flower structures.: This study explores the use of femtosecond laser techniques for structuring silver vanadium oxide into flower-like formations, potentially useful for enhancing the material′s applicability in various optical applications (Sun et al., 2019).

Analysis Note

Assay (redox titration): ≥ 99.0 %
Chloride (Cl): ≤ 0.002 %
Phosphate (PO₄): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.05 %
Fe (Iron): ≤ 0.003 %
Pb (Lead): ≤ 0.003 %
Corresponds to Ph Eur

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
01-478010028398128
assay

≥99%

assay

≥99.0%

assay

≥99.0% (RT)

assay

≥99.0%

Quality Level

100

Quality Level

-

Quality Level

200

Quality Level

200

agency

reag. Ph. Eur.

agency

-

agency

-

agency

-

form

solid

form

solid

form

powder or crystals

form

powder

pH

7 (20 °C, 5.1 g/L in H2O)

pH

-

pH

7 (20 °C, 5.1 g/L)

pH

7 (20 °C, 5.1 g/L)

solubility

5.1 g/L

solubility

-

solubility

-

solubility

NH4OH: passes test


signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - STOT RE 1 Inhalation

target_organs

Respiratory Tract

Storage Class

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters