Skip to Content
MilliporeSigma
All Photos(2)

Documents

1.04936

Supelco

Potassium chloride

for analysis EMSURE®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-211-8
NACRES:
NA.21

Quality Level

product line

EMSURE®

Assay

≥99.5% (silver nitrate titration)

form

solid

potency

2600 mg/kg LD50, oral (Rat)

impurities

≤0.01% In water insoluble matter

pH

5.5-8.0 (25 °C, 50 g/L in H2O)

bp

1413 °C/1013 hPa

mp

770 °C (lit.)

solubility

347 g/L

density

1.98 g/mL at 25 °C (lit.)

bulk density

1000 kg/m3

anion traces

bromide (Br-): ≤0.05%
iodide (I-): ≤0.002%
phosphate (PO43-): ≤0.0005%
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.001%
Ca: ≤0.001%
Fe: ≤0.0003%
Mg: ≤0.002%
N: ≤0.001%
Na: ≤0.02%
heavy metals (as Pb): ≤0.0005%

storage temp.

2-30°C

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.

Analysis Note

Assay (argentometric): ≥ 99.5 %
In water insoluble matter: ≤ 0.01 %
pH-value (5 %; water): 5.5 - 8.0
Bromide (Br): ≤ 0.05 %
Iodide (I): ≤ 0.002 %
Phosphate (PO₄): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0005 %
Ba (Barium): ≤ 0.001 %
Ca (Calcium): ≤ 0.001 %
Fe (Iron): ≤ 0.0003 %
Mg (Magnesium): ≤ 0.002 %
Na (Sodium): ≤ 0.02 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

EMSURE® Grade Inorganic Solvents and Reagents offer exceptional purity and quality control, ensuring reliable, compliant results for regulated analyses.

EMSURE® Grade Inorganic Solvents and Reagents offer exceptional purity and quality control, ensuring reliable, compliant results for regulated analyses.

EMSURE® Grade Inorganic Solvents and Reagents offer exceptional purity and quality control, ensuring reliable, compliant results for regulated analyses.

EMSURE® Grade Inorganic Solvents and Reagents offer exceptional purity and quality control, ensuring reliable, compliant results for regulated analyses.

Protocols

A complete SEC-UV workflow for quantifying monoclonal antibody fragments, monomers, and aggregates, including procedure for a system suitability test and heat stressing of antibodies.

A complete SEC-UV workflow for quantifying monoclonal antibody fragments, monomers, and aggregates, including procedure for a system suitability test and heat stressing of antibodies.

A complete SEC-UV workflow for quantifying monoclonal antibody fragments, monomers, and aggregates, including procedure for a system suitability test and heat stressing of antibodies.

A complete SEC-UV workflow for quantifying monoclonal antibody fragments, monomers, and aggregates, including procedure for a system suitability test and heat stressing of antibodies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service