Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.04340

Supelco

Isohexane

≥99.8% (GC), suitable for gas chromatography (GC) ECD and FID, SupraSolv®

Synonym(s):

Isohexane, 2-Methyl pentane, iso-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14
CAS Number:
Molecular Weight:
86.18
MDL number:
UNSPSC Code:
12191502
EC Index Number:
295-570-2
NACRES:
NA.03
Assay:
≥99.8% (GC)
technique(s):
gas chromatography (GC): suitable
bp:
53-63 °C/1013 hPa
vapor pressure:
160-190 hPa ( 20 °C)

Product Name

Isohexane, for gas chromatography ECD and FID SupraSolv®

vapor pressure

160-190 hPa ( 20 °C)

Quality Level

product line

SupraSolv®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

260 °C

potency

>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.0-7.4 % (v/v)

technique(s)

gas chromatography (GC): suitable

kinematic viscosity

0.5 cSt(20 °C)

bp

53-63 °C/1013 hPa

mp

<-50 °C

transition temp

flash point -21 °C

solubility

0.02 g/L

density

0.66 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(CCC)CC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

General description

SupraSolv ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Application



  • Enantiomeric determination of tramadol and its main metabolite O-desmethyltramadol in human plasma by liquid chromatography-tandem mass spectrometry.: Explores the role of isohexane in enhancing the enantioselective analysis of tramadol, which is critical for therapeutic drug monitoring and pharmacokinetic studies (Ceccato et al., 2000).


Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-5.8 °F

Flash Point(C)

-21 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service