Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

1.04333

Supelco

Isohexane

≥95.0% (C₆H₁₄-isomeres, GC) or analysis EMSURE®

Synonym(s):

Isohexane, 2-Methyl pentane, iso-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14
CAS Number:
Molecular Weight:
86.18
MDL number:
UNSPSC Code:
12191502
EC Index Number:
295-570-2
NACRES:
NA.21
grade:
reagent grade
bp:
53-63 °C/1013 hPa
vapor pressure:
160-190 hPa ( 20 °C)

Product Name

Isohexane, for analysis EMSURE®

grade

reagent grade

Quality Level

vapor pressure

160-190 hPa ( 20 °C)

product line

EMSURE®

Assay

≥95.0% (C<SUB>6</SUB>H<SUB>14</SUB>-isomeres, GC)

form

liquid

autoignition temp.

260 °C

potency

>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.0-7.4 % (v/v)

dilution

(for analytical testing)

impurities

≤0.0001% Thiophene
≤0.0003 meq/g Acidity
≤0.005% Sulfur compounds (as S)
≤0.01% Aromatics (as benzene)
≤0.01% Water
≤5% n-Hexane (GC)

evapn. residue

≤10 mg/L

kinematic viscosity

0.5 cSt(20 °C)

bp

53-63 °C/1013 hPa

mp

<-50 °C

transition temp

flash point -21 °C

density

0.66 g/cm3 at 20 °C

cation traces

Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%

storage temp.

2-30°C

SMILES string

C(CCC)CC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Linkage

Discover EMSURE - to be more than sure!

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-5.8 °F

Flash Point(C)

-21 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service