Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
NACRES:
NA.03
EC Index Number:
200-835-2
UNSPSC Code:
12191502
MDL number:
Beilstein/REAXYS Number:
741857
Assay:
≥99.8% (GC)
Technique(s):
gas chromatography (GC): suitable
Bp:
81-82 °C (lit.)
Vapor pressure:
72.8 mmHg ( 20 °C), 97 hPa ( 20 °C)
Pricing and availability is not currently available.
Product Name
Acetonitrile, for gas chromatography ECD and FID SupraSolv®
Quality Segment
vapor density
1.41 (vs air)
vapor pressure
72.8 mmHg ( 20 °C), 97 hPa ( 20 °C)
product line
SupraSolv®
assay
≥99.8% (GC)
form
liquid
autoignition temp.
524 °C, 973 °F
expl. lim.
16 %
technique(s)
gas chromatography (GC): suitable
refractive index
n20/D 1.344 (lit.)
bp
81-82 °C (lit.)
mp
−45 °C (lit.)
transition temp
flash point 2 °C
density
0.786 g/mL at 25 °C (lit.)
format
neat
storage temp.
2-30°C
SMILES string
CC#N
InChI
1S/C2H3N/c1-2-3/h1H3
InChI key
WEVYAHXRMPXWCK-UHFFFAOYSA-N
General description
SupraSolv® ECD and FID is a high-purity solvent grade specifically formulated for sample preparation in gas chromatography with electron capture (ECD) and flame ionization (FID) detection.ECD and FID impose distinct and stringent solvent purity requirements. The electron capture detector is exquisitely sensitive to electronegative species — particularly halogenated compounds — at sub-nanogram concentrations, while the flame ionization detector responds to any carbon-containing organic impurity undergoing combustion. SupraSolv® ECD and FID solvents are therefore subjected to rigorous purification and detector-specific testing protocols to ensure that intrinsic solvent impurities remain below the respective detection thresholds of both detector types simultaneously.
Application
These solvents are routinely employed in PCB and organochlorine determination in water and soil, pesticide residue analysis in food and environmental matrices, dioxin screening, and hydrocarbon index measurements — applications where detector sensitivity demands the highest attainable solvent purity to ensure method selectivity and regulatory compliance.
Features and Benefits
• Dual-detector validation for both ECD and FID within a single solvent grade
• Detector-specific impurity control, targeting the precise analyte classes most detrimental to ECD (halogenated compounds) and FID (hydrocarbons and organic residues) performance, resp.
• Widest specified retention time range, guaranteeing a clean chromatographic baseline from early- to late-eluting analytes
• Batch-to-batch consistency, ensuring long-term reproducibility across routine and regulatory analytical workflows
• Detector-specific impurity control, targeting the precise analyte classes most detrimental to ECD (halogenated compounds) and FID (hydrocarbons and organic residues) performance, resp.
• Widest specified retention time range, guaranteeing a clean chromatographic baseline from early- to late-eluting analytes
• Batch-to-batch consistency, ensuring long-term reproducibility across routine and regulatory analytical workflows
Analysis Note
SupraSolv® ECD and FID solvents are tested and certified against stringent detector-specific purity thresholds across defined chromatographic retention time windows, including:
- GC-ECD (early retention window): ≤ 1 ng/mL relative to tetrachloromethane
- GC-ECD (late retention window): ≤ 2 pg/mL relative to lindane
- GC-FID: ≤ 2 ng/mL relative to n-tetradecane
Other Notes
Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.
Legal Information
SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under Acetonitrile
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
flash_point_f
35.6 °F - closed cup
flash_point_c
2.0 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

