Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.02817

Supelco

Cyclohexane

for gas chromatography ECD and FID SupraSolv®

Synonym(s):

Hexahydrobenzene, Hexamethylene, Naphthene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900225
MDL number:
UNSPSC Code:
12191504
EC Index Number:
203-806-2
NACRES:
NA.03
Assay:
≥99.8% (GC)
technique(s):
gas chromatography (GC): suitable
bp:
80.7 °C (lit.)
vapor pressure:
168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

vapor density

2.9 (vs air)

Quality Level

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

product line

SupraSolv®

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

500 °F

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

9 %

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.426 (lit.)

kinematic viscosity

1.26 cSt(20 °C)

bp

80.7 °C (lit.)

mp

4-7 °C (lit.)

transition temp

flash point -18 °C

solubility

0.05 g/L

density

0.779 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

InChI key

XDTMQSROBMDMFD-UHFFFAOYSA-N

General description

SupraSolv ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service