Skip to Content
MilliporeSigma

1.08597

SAFC

Hydrogen peroxide

30%, stabilized, EMPROVE® ESSENTIAL, Ph. Eur., BP, USP

Pharma Manufacturing

Synonym(s):

Hydrogen peroxide solution, Perhydrol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2O2
CAS Number:
Molecular Weight:
34.01
Beilstein:
3587191
MDL number:
UNSPSC Code:
12352304

Agency

BP
Ph. Eur.
USP

Quality Level

vapor pressure

18 hPa ( 20 °C)
23.3 mmHg ( 30 °C)

product line

EMPROVE® ESSENTIAL

form

liquid

concentration

30%

pH

2-4 (20 °C in H2O)

bp

107 °C/1013 hPa

mp

-26 °C

density

1.11 g/cm3 at 20 °C

application(s)

liquid formulation
pharma/biopharma processes
pharmaceutical
semi-solid formulation

storage temp.

2-25°C

SMILES string

OO

InChI

1S/H2O2/c1-2/h1-2H

InChI key

MHAJPDPJQMAIIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Hydrogen peroxide is commonly used as preservative in pharmaceutical formulations due to its potent and broad-spectrum antimicrobial effects.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

not combustible

Flash Point(C)

not combustible


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christoph Plieth
ACS omega, 4(2), 3268-3279 (2019-08-29)
The peroxidation of luminol yields bright luminescence when the reaction is catalyzed by heme proteins. However, an excess of peroxide leads to less light and altered luminescence kinetics, an effect commonly referred to as "suicide inactivation". The aim of this
Christoph Plieth
ACS omega, 3(9), 12295-12303 (2019-08-29)
Iron porphyrin catalysts of the luminol reaction (horseradish peroxidase, hemoglobin, cytochrome c, and hemin) interact with diverse reducing compounds. Here, it is demonstrated how the chemiluminescence yield is modulated by such interactions. The compounds accepted as substrates protect the catalyst

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service