Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

I-038

Supelco

Isobutyryl fentanyl hydrochloride solution

100 μg/mL in methanol (as a free base), certified reference material, ampule of 0.5 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30N2O · HCl
CAS Number:
Molecular Weight:
386.96
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 0.5 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as a free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CC(C)C(N(C1=CC=CC=C1)C2CCN(CCC3=CC=CC=C3)CC2)=O.Cl

InChI

1S/C23H30N2O.ClH/c1-19(2)23(26)25(21-11-7-4-8-12-21)22-14-17-24(18-15-22)16-13-20-9-5-3-6-10-20;/h3-12,19,22H,13-18H2,1-2H3;1H

InChI key

RAMJZGOMHMJBHA-UHFFFAOYSA-N

General description

A certified Snap-N-Spike® solution suitable for use as starting material in calibrators and controls for isobutyryl fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, prescription monitoring, or forensic analysis applications. Isobutyryl fentanyl is a fentanyl analog and sold on the illicit market as a designer drug or in combination with heroin.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service