Skip to Content
MilliporeSigma
All Photos(1)

Documents

F-049

Supelco

p-Fluorofentanyl solution

100 μg/mL in methanol, certified reference material, ampule of 0.5 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H27FN2O
CAS Number:
Molecular Weight:
354.46
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 0.5 mL

manufacturer/tradename

Cerilliant®

drug control

estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

InChI

1S/C22H27FN2O/c1-2-22(26)25(20-10-8-19(23)9-11-20)21-13-16-24(17-14-21)15-12-18-6-4-3-5-7-18/h3-11,21H,2,12-17H2,1H3

InChI key

KXUBAVLIJFTASZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

para-Fluorofentanyl is an opioid analgesic being an analogue of fentanyl developed by Janssen Pharmaceutica in the 1960s. para-Fluorofentanyl potently activates the human μ; opioid receptor and is abused recreationally. A certified Snap-N-Spike® solution suitable for use as starting material in calibrators and controls for para-Fluorofentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing or forensic analysis applications.

Application


  • Quantitative analysis of synthetic opioids: A study presents a quantitative LC-MS/MS method for analyzing p-Fluorofentanyl along with Xylazine, Fentanyl, and related compounds in postmortem blood, aiding forensic and pharmacological research in opioid toxicity (Truver et al., 2024).

  • Detection in biological specimens: Research on the detection of Fentanyl, synthetic opioids including p-Fluorofentanyl, and ketamine in hair samples from American and Italian populations provides critical data for understanding patterns of opioid abuse and assisting in drug monitoring programs (Salomone et al., 2023).

  • Innovative forensic techniques: A study introduces the use of pressure-sensitive adhesives and paper spray-mass spectrometry for the efficient collection and analysis of p-Fluorofentanyl and related compounds from shipping materials, highlighting advancements in forensic methodologies (Prunty et al., 2023).

  • Pharmacological profile and toxicological assessment: An exploration of fentanyl analog structure-activity relationships including p-Fluorofentanyl examines the determinants of their potencies for antinociception versus respiratory depression, crucial for developing safer analgesic agents (Varshneya et al., 2023).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service