Skip to Content
MilliporeSigma
All Photos(1)

Documents

F-048

Supelco

p-Fluorobutyryl fentanyl (PFBF) solution

100 μg/mL in methanol (), certified reference material, ampule of 0.5 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H29FN2O
CAS Number:
Molecular Weight:
368.49
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 0.5 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (
)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

InChI

1S/C23H29FN2O/c1-2-6-23(27)26(21-11-9-20(24)10-12-21)22-14-17-25(18-15-22)16-13-19-7-4-3-5-8-19/h3-5,7-12,22H,2,6,13-18H2,1H3

InChI key

QZFMCYUBPSLOBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A certified Snap-N-Spike®; solution suitable for use as starting material in calibrators and controls in para-Fluorobutyryl fentanyl LC/MS or GC/MS testing methods for clinical toxicology, urine drug testing or forensic analysis applications. para-Fluorobutyryl fentanyl, also known as PFBF and 4-FBF, is a designer drug and fentanyl analog that is abused recreationally.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service