Skip to Content
MilliporeSigma

A-129

Acetyl fentanyl solution

50 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H26N2O
CAS Number:
Molecular Weight:
322.44
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal); Kábítószer / Narcotic Drug (Hungary), 78/2022. (XII. 28.) BM rendelet

concentration

50 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C)N(C1CCN(CCC2=CC=CC=C2)CC1)C3=CC=CC=C3

InChI

1S/C21H26N2O/c1-18(24)23(20-10-6-3-7-11-20)21-13-16-22(17-14-21)15-12-19-8-4-2-5-9-19/h2-11,21H,12-17H2,1H3

InChI key

FYIUUQUPOKIKNI-UHFFFAOYSA-N

General description

A certified Snap-N-Spike® solution suitable for use as starting material in calibrators and controls for fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, prescription monitoring, or forensic analysis applications. Acetyl fentanyl is a designer drug and fentanyl analog with a potency 40 times greater than heroin and 80 times greater than morphine. This Certified Spiking Solution® is supplied in a convenient, quantitative, US DEA-exempt solution format and with TK #s for Canadian customers.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.


Still not finding the right product?

Explore all of our products under Acetyl fentanyl solution


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library