General description
Photo KitAlysis Operating Instructions
Features:
- Designed and tested by synthetic chemists.
- Controller provides repeatable milliamp selection for photon intensity (sold seperately)
- 527 nm wavelength
- 0-30 mA variable LED output
- Non-magnetic LED base
- Chemically resistant LED cover
- PTFE coated cabling
Photo Kitalysis Starter Kit required for operation (sold separately). Best when used with KitAlysis Benchtop Inertion Box (sold separately).
Legal Information
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
Articles
PhotoKitAlysis Reaction Kit aids in cross-coupling reactions, offering pre-weighed components for optimization.
PhotoKitAlysis Reaction Kit aids in cross-coupling reactions, offering pre-weighed components for optimization.
PhotoKitAlysis Reaction Kit aids in cross-coupling reactions, offering pre-weighed components for optimization.
PhotoKitAlysis Reaction Kit aids in cross-coupling reactions, offering pre-weighed components for optimization.
Protocols
Materials and procedure Step-by-Step Guide for Photocatalysis High-Throughput Reaction Screening Kit
Materials and procedure Step-by-Step Guide for Photocatalysis High-Throughput Reaction Screening Kit
Materials and procedure Step-by-Step Guide for Photocatalysis High-Throughput Reaction Screening Kit
Materials and procedure Step-by-Step Guide for Photocatalysis High-Throughput Reaction Screening Kit