Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

KITALYSIS-BOR

Sigma-Aldrich

KitAlysis High-Throughput Miyaura Borylation Reaction Screening Kit-Pack of 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161600
NACRES:
NA.21

reaction suitability

reagent type: catalyst
core: palladium
reaction type: Miyaura Borylation Reaction

functional group

phosphine

General description

The KitAlysis High-Throughput Miyaura Borylation Reaction Screening Kit enables chemists to quickly and efficiently screen reaction conditions for Pd catalyzed Miyaura borylation reactions. Quick identification of optimal conditions allows faster scale-up of the desired synthetic transformation. All required chemicals come pre-weighed for ease of use. A step-by-step user guide and experimental excel sheets are available for download to facilitate the process.

Application

Step-by-step user guides and a downloadable experimental excel sheets are provided in the below hyperlinks.
Step-by-Step Guide for Miyaura Borylation Reaction Screening Kit
Downloadable Experiment excel sheets

Each kit contains 2 customizable screening sets that include the following components:

  • 6 x 4 pre-weighed catalysts in glass vials loaded with stir bars and topped with cap mat.
  • 4 x 4 mL reaction vials with pre-weighed KOAc
  • Bis(pinacolato)diboron (473294), tetrahydroxydiboron (754242), and pinacol (221171)
  • 1 (2 mL) ampule each of toluene, THF, n-butanol, DMA, MeOH, MeCN, ethylene glycol and DIPEA.
  • 1 NEW KitAlysis 24-Well Reaction Block Replacement Film.
  • 4 stir bars for substrate stock solution vials.
  • 1 internal standard.
Monitor reaction progress via TLC-MS by following this protocol

Best when used with KitAlysis Benchtop Inertion Box and 24-well Reaction Block (sold separately).

Features and Benefits

  • Designed and tested by synthetic chemists.
  • Easily run 24-microscale reactions in parallel.
  • 10 μmol reactions with 1 μmol catalyst, 3 equiv. base, 0.1 M concentration.
  • Customizable Excel Template for quick calculations and easy incorporation into electronic laboratory notebooks.

Legal Information

KitAlysis is a trademark of Sigma-Aldrich Co. LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 764175P(Cy3) Pd G3, 97%SDS

  • 791733Potassium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%SDS

  • 804274XPhos Pd G4, 95%SDS

  • 804282SPhos Pd G4SDS

  • 804290RuPhos Pd G4SDS

  • 804975DTBPF-Pd-G3SDS

  • 900349cataCXium A Pd G4SDS

  • 900522Toluene, anhydrous, ZerO2®, 99.8%SDS

  • 900631Ethylene glycol, anhydrous, ZerO2®, 99.8%SDS

  • 900634N,N-Dimethylacetamide, anhydrous, ZerO2®, 99.8%SDS

  • 900636Tetrahydrofuran, anhydrous, inhibitor-free, ZerO2®, ≥99.9%SDS

  • 9006391-Butanol, anhydrous, ZerO2®, 99.8%SDS

  • 900641Methanol, anhydrous, ZerO2®, 99.8%SDS

  • 900644Acetonitrile, anhydrous, ZerO2®, 99.8%SDS

  • 900690N,N-Diisopropylethylamine, purified by redistillation, ZerO2®, 99.5%SDS

  • 221171Pinacol, 98%SDS

  • 473294Bis(pinacolato)diboron, 99%SDS

  • 754242Tetrahydroxydiboron, 95%SDS

  • B34656Biphenyl, ReagentPlus®, 99.5%SDS

  • Z118850Spinbar® magnetic stirring fleas, red, L 10 mm, diam. 3 mmSDS

  • Z742110KitAlysis 24-well reaction block replacement filmSDS

See All (21)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes,Central nervous system, Kidney, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

KitAlysis High-Throughput Screening Kits provide solution to efficiently identify or optimize suitable catalytic reaction conditions. Chemist can rapidly run 24 unique micro scale reactions in parallel with tailored conditions.

Protocols

Catalyst screening kits, KitAlysis™, and TLC-MS analysis enable high-throughput synthesis and analysis for efficient reactions.

Catalyst screening kits, KitAlysis™, and TLC-MS analysis enable high-throughput synthesis and analysis for efficient reactions.

Catalyst screening kits, KitAlysis™, and TLC-MS analysis enable high-throughput synthesis and analysis for efficient reactions.

Catalyst screening kits, KitAlysis™, and TLC-MS analysis enable high-throughput synthesis and analysis for efficient reactions.

Related Content

Chemical reaction design and optimization in organic synthesis aims to precisely alter reaction parameters to achieve optimum results in finding pathways to target molecules.

Chemical reaction design and optimization in organic synthesis aims to precisely alter reaction parameters to achieve optimum results in finding pathways to target molecules.

Chemical reaction design and optimization in organic synthesis aims to precisely alter reaction parameters to achieve optimum results in finding pathways to target molecules.

Chemical reaction design and optimization in organic synthesis aims to precisely alter reaction parameters to achieve optimum results in finding pathways to target molecules.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service