Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z742680

Sigma-Aldrich

SynLED Parallel Photoreactor

Synonym(s):

parallel photoreactor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352300

AC/DC input

, universal plug set (included)
100 - 240 V AC, 50/60 Hz

feature

capacity 16  vial(s)

reaction suitability

reagent type: catalyst
reaction type: Photocatalysis

parameter

465-470 nm spectral range

General description

This newly designed photoreactor enables simple small-scale photocatalysis reaction screening ensuring high levels of consistency across reactions and between runs.

Features and Benefits

  • Bottom-lit LEDs (465-470 nm) across a 4x4 reaction block array provides consistent light intensity (130-140 lm) and angle (45°)
  • Built-in cooling fan provides consistent temperature to each parallel reaction
  • Uses 1-2 dram scintillation vials or microwave vials (O.D. of 1.7 cm or less)
  • Power supply is wall plug power supply 700mA 12 W
  • Compatible with IKA magnetic stirrer with round plate (Z645052)
  • Compatible with liquid scintillation vials (8 mL Wheaton, Z188719)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This review provides a summary of recent approaches in developing green methods for visible-light-mediated reactions that operate under mild conditions and avoid the need for toxic transition-metal catalysts or harsh reaction conditions.

This review provides a summary of recent approaches in developing green methods for visible-light-mediated reactions that operate under mild conditions and avoid the need for toxic transition-metal catalysts or harsh reaction conditions.

This review provides a summary of recent approaches in developing green methods for visible-light-mediated reactions that operate under mild conditions and avoid the need for toxic transition-metal catalysts or harsh reaction conditions.

This review provides a summary of recent approaches in developing green methods for visible-light-mediated reactions that operate under mild conditions and avoid the need for toxic transition-metal catalysts or harsh reaction conditions.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service