Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z744032

Sigma-Aldrich

420nm Light Source

Synonym(s):

Penn PhD Photoreactor M2 light

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161600
NACRES:
NA.22

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

General description

The 420nm LED Light Module is used with the Penn PhD Photoreactor M2 for chemists and researchers to accelerate chemical reactions using photoredox catalysis.

Features and Benefits of Penn PhD Photoreactor M2
  • Modular design allows for use with a variety of wavelengths 365nm – 450nm
  • 360 degree reflective environment maximizes surface area photon capture
  • Light shield interlock prevents user exposure to harmful light rays
  • Interactive touch screen controls reaction parameters
  • Intertek ETL, CE, and CB approved
  • User defined parameters including temperature, light intensity, fan speed and stirring
  • Auto stop, pause and reset options
  • Supports vial sizes gc, 4, 8, 20, 40 ml
  • Temp feedback using a k-type thermocouple


Photocatalysis Technology Spotlight

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Effective Csp2- and Csp-hybridized coupling reactions established for catalysis, expanding to multi-step Csp3-coupling.

Effective Csp2- and Csp-hybridized coupling reactions established for catalysis, expanding to multi-step Csp3-coupling.

Photoredox catalysis is a powerful synthetic methodology to form challenging covalent bonds using light irradiation. It is effective for light-driven polymer and small molecule synthesis.

Effective Csp2- and Csp-hybridized coupling reactions established for catalysis, expanding to multi-step Csp3-coupling.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service