Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

Z742107

Sigma-Aldrich

KitAlysis 24-well Reaction Block and Screwdriver Set

Synonym(s):

Reaction block

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.51

reaction suitability

reaction type: Cross Couplings

Quality Level

General description

Use in combination with the KitAlysis Bench Top Inertion Box (Z742064) or a glove box/glove bag to provide inert atmosphere for kit set-up.
Designed to be used with KitAlysis High-Throughput Screening Kits.

Components:
  • 24-Well Reaction Block
  • Torque Screwdriver
  • Small screwdriver to easily remove torqued screws after reaction is complete.
  • 10 Reaction Block Replacement Screws

Features and Benefits

  • Torque screwdriver specially designed to seal the block for minimal solvent loss while heating at elevated temperatures.
  • Torque setting prevents over-tightening and stripping of screws.
  • Includes small screwdriver to easliy remove torqued screws after reaction is complete.

Legal Information

KitAlysis is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Solutions & slurries to make: Go to the online user set-up page to enter molecular weights into the downloadable excel file.

See All

Related Content

Solutions & slurries to make: Go to the online user set-up page to enter molecular weights into the downloadable excel file.

Solutions & slurries to make: Go to the online user set-up page to enter molecular weights into the downloadable excel file.

Solutions & slurries to make: Go to the online user set-up page to enter molecular weights into the downloadable excel file.

Solutions & slurries to make: Go to the online user set-up page to enter molecular weights into the downloadable excel file.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service