Documents
Z680273
Titanium tweezers
6 piece kit
Synonym(s):
Precision Tweezers, Titanium Forceps
About This Item
UNSPSC Code:
27112105
NACRES:
MA.01
manufacturer/tradename
Aven
General description
This set consists of our most popular titanium tweezers in a handy pouch. Tweezers included in the set are Styles 2A, 3c, OO, AA, 5 and 7.
Titanium tweezers do not shed particles like stainless steel tweezers. Titanium is light weight, high strength, non-magnetic material.
  • Lightweight (high strength to weight ratio)
  • Zero magnetic retentively
  • Temperature resistant to 800 °F
  • Chemical and corrosion resistant
  • Non-solder adhesive
  • Bio-compatible (maintain cell integrity, no inflammatory response)
Typical application: handling of sensitive components in cleaning/chemical processes at high temperatures, histology, biology, forensics, electron microscopy etc.
Visit the Document Library
Customers Also Viewed
Slide 1 of 9
1 of 9